Orders LinkContactUsLinkComponent
Pedestals, solid, SC 40-series
Pedestals, solid, SC 40-series
Nome del fornitore:   HAMMERBACHER
Pedestals, solid, SC 40-series
Nome del fornitore:  HAMMERBACHER
Certificates
Pedestals, solid, SC 40-series
Pedestals, solid, SC 40-series
32 varianti
Product Details & Documents
Standing container with 2 ORGA drawers, one full-extension suspension file drawer and one utility drawer. The pedestal height is the same as the standard desk height so by placing the pedestal adjacent to a desk the work surface can be extended.
  • Central locking with bidirectional drawer stop
  • ACSA material feed tray and ACTR separating strip optionally available as accessories
  • Various colours and handles to choose from
  • Height adjustment: 720 to 760 mm using levelling feet
Material: Melamine resin coated plate in accordance with DIN 68765, emission class E 1.
Pen tray: top utility drawer, sliding on plastic guide.
Drawer: ORGA plastic drawer on metal drawer runners, 80% extension, self-closing, load capacity: 25 kg.
Suspension drawer: ORGA plastic drawer, on metal drawer runners, full extension, with two removable frames for holding suspension files (in parallel or diagonally); load capacity: 25 kg.
Delivery information: The unit is delivered completely assembled; only handles and rollers need to be attached.
Product Family Options
Informazioni sul prodotto
  • L×P×A
    Disponibilità
    Price
  • HAMBVSC40/3/3/BM
    Standard container, iron handle, maple
    Standard container, iron handle, maple
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/3/3/CE
    Standard container, chrome handle, maple
    Standard container, chrome handle, maple
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/3/3/KR
    Standard container, cross handle, maple
    Standard container, cross handle, maple
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/3/3/NU
    Standard container, nut handle, maple
    Standard container, nut handle, maple
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/3/3/RE
    Standard container, railing handle, maple
    Standard container, railing handle, maple
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/3/3/SG
    Standard container, strip handle, maple
    Standard container, strip handle, maple
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/3/3/WE
    Standard container, wave handle, maple
    Standard container, wave handle, maple
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/3
    Combination cupboard, maple
    Combination cupboard, maple
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/5/5/BM
    Standard container, iron handle, grey
    Standard container, iron handle, grey
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/5/5/CE
    Standard container, chrome handle, grey
    Standard container, chrome handle, grey
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/5/5/KR
    Standard container, cross handle, grey
    Standard container, cross handle, grey
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/5/5/NU
    Standard container, nut handle, grey
    Standard container, nut handle, grey
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/5/5/RE
    Standard container, railing handle, grey
    Standard container, railing handle, grey
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/5/5/SG
    Standard container, strip handle, grey
    Standard container, strip handle, grey
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/5/5/WE
    Standard container, wave handle, grey
    Standard container, wave handle, grey
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/5
    Combination cupboard, grey
    Combination cupboard, grey
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/6/6/BM
    Standard container, iron handle, beech
    Standard container, iron handle, beech
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/6/6/CE
    Standard container, chrome handle, beech
    Standard container, chrome handle, beech
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/6/6/KR
    Standard container, cross handle, beech
    Standard container, cross handle, beech
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/6/6/NU
    Standard container, nut handle, beech
    Standard container, nut handle, beech
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/6/6/RE
    Standard container, railing handle, beech
    Standard container, railing handle, beech
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/6/6/SG
    Standard container, strip handle, beech
    Standard container, strip handle, beech
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/6/6/WE
    Standard container, wave handle, beech
    Standard container, wave handle, beech
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/6
    Combination cupboard, beech
    Combination cupboard, beech
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/W/W/BM
    Standard container, iron handle, white
    Standard container, iron handle, white
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/W/W/CE
    Standard container, chrome handle, white
    Standard container, chrome handle, white
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/W/W/KR
    Standard container, cross handle, white
    Standard container, cross handle, white
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/W/W/NU
    Standard container, nut handle, white
    Standard container, nut handle, white
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/W/W/RE
    Standard container, railing handle, white
    Standard container, railing handle, white
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/W/W/SG
    Standard container, strip handle, white
    Standard container, strip handle, white
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/W/W/WE
    Standard container, wave handle, white
    Standard container, wave handle, white
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HAMBVSC40/Z/W/WE
    Standard container, wave handle, plum
    Standard container, wave handle, plum
    428×800×720 - 760 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Dividers ACTR, colour: grey, suitable for ORGA drawers, 4 x A 4, 2 x A 5, 2 x A 6
    Dividers ACTR, colour: grey, suitable for ORGA drawers, 4 x A 4, 2 x A 5, 2 x A 6
    HAMBVACTR
    Log in per disponibilità
    1 * 1 ST
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Set of inclined shelves ASCA, colour: gray 32.7 x 21.2 x 3.3 (WxDxH), suitable for ORGA drawers
    Set of inclined shelves ASCA, colour: gray 32.7 x 21.2 x 3.3 (WxDxH), suitable for ORGA drawers
    HAMBVACSA
    Log in per disponibilità
    1 * 5 ST