Cleanroom trousers, elasticated
Informazioni su questo articolo
These trousers are made of XA32 fabric consisting of 99% polyester and 1% conductive fibres. The fibres are continuous filament fibres, which means that the garments do not emit particles or fibres. The carbon filaments produce good static discharge, thus preventing static charging and minimising triboelectric attraction of particles and fibres. This makes the garments very suitable for activities such as vehicle spraying.
- Tracksuit-style
- Elasticated waistband with drawstring
- Open side legs
- Inside key pocket
Weight: 125 g/m²
6 varianti
Product Details & Documents
These trousers are made of XA32 fabric consisting of 99% polyester and 1% conductive fibres. The fibres are continuous filament fibres, which means that the garments do not emit particles or fibres. The carbon filaments produce good static discharge, thus preventing static charging and minimising triboelectric attraction of particles and fibres. This makes the garments very suitable for activities such as vehicle spraying.
- Tracksuit-style
- Elasticated waistband with drawstring
- Open side legs
- Inside key pocket
Weight: 125 g/m²
Documents
Specifications for Product Family
Specifiche
- Catalog No:
- KWIN100631410XS
- KWIN100631410S
- KWIN100631410M
- KWIN100631410L
- KWIN100631410XL
- KWIN1006314102XL
- Modello:
- Elasticated waist
- Elasticated waist
- Elasticated waist
- Elasticated waist
- Elasticated waist
- Elasticated waist
- Applicazioni per abiti:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Risk area:
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Tessuto:
- 99% polyester and 1% conductive fibre
- 99% polyester and 1% conductive fibre
- 99% polyester and 1% conductive fibre
- 99% polyester and 1% conductive fibre
- 99% polyester and 1% conductive fibre
- 99% polyester and 1% conductive fibre
- Peso:
- 125 g/m²
- 125 g/m²
- 125 g/m²
- 125 g/m²
- 125 g/m²
- 125 g/m²
Specifiche
Product Family Options
Informazioni sul prodotto
- ColoreTaglia dell'abbigliamentoDisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.KWIN100631410XSModelloElasticated waistApplicazioni per abitiCleanroomRisk areaAnti-staticGenereUnisexSleeve or leg lengthLongTessuto99% polyester and 1% conductive fibrePeso125 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100631410SModelloElasticated waistApplicazioni per abitiCleanroomRisk areaAnti-staticGenereUnisexSleeve or leg lengthLongTessuto99% polyester and 1% conductive fibrePeso125 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100631410MModelloElasticated waistApplicazioni per abitiCleanroomRisk areaAnti-staticGenereUnisexSleeve or leg lengthLongTessuto99% polyester and 1% conductive fibrePeso125 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100631410LModelloElasticated waistApplicazioni per abitiCleanroomRisk areaAnti-staticGenereUnisexSleeve or leg lengthLongTessuto99% polyester and 1% conductive fibrePeso125 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100631410XLModelloElasticated waistApplicazioni per abitiCleanroomRisk areaAnti-staticGenereUnisexSleeve or leg lengthLongTessuto99% polyester and 1% conductive fibrePeso125 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN1006314102XLModelloElasticated waistApplicazioni per abitiCleanroomRisk areaAnti-staticGenereUnisexSleeve or leg lengthLongTessuto99% polyester and 1% conductive fibrePeso125 g/m²