Orders LinkContactUsLinkComponent
Optical components for optical bench
Optical components for optical bench
Nome del fornitore:   DIDALAB
Optical components for optical bench
Nome del fornitore:  DIDALAB
Certificates
Optical components for optical bench
Optical components for optical bench
11 varianti
Product Details & Documents
Product Family Options
Informazioni sul prodotto
  • Disponibilità
    Price
  • DIDAPOD066700
    Slits on glass plate (Ø 40 mm), 7 slits and wires, widths : 30, 40, 60, 80, 100, 150, 200 µm
    Slits on glass plate (Ø 40 mm), 7 slits and wires, widths : 30, 40, 60, 80, 100, 150, 200 µm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 35.00
    SFr. 35.00
    1 * 1 ST
       SFr. 35.00
  • DIDAPOD066710
    Young's slits on a glass plate (Ø 40 mm), 3 double slits with distance between slits 200, 400, 500 µm
    Young's slits on a glass plate (Ø 40 mm), 3 double slits with distance between slits 200, 400, 500 µm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 35.00
    SFr. 35.00
    1 * 1 ST
       SFr. 35.00
  • DIDAPOD068282
    Direct vision prism with internal Amici prism
    Direct vision prism with internal Amici prism
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • DIDAPOD013020
    Fresnel mirrors, one mirror is fixed, the other one has a tilting adjustment device, mirror size 45×40 mm
    Fresnel mirrors, one mirror is fixed, the other one has a tilting adjustment device, mirror size 45×40 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 131.00
    SFr. 131.00
    1 * 1 ST
       SFr. 131.00
  • DIDAPOD060210
    Beam splitter, enables the incident beam to be divided into two emergent beams of identical light intensity, variable angle of emergent beam
    Beam splitter, enables the incident beam to be divided into two emergent beams of identical light intensity, variable angle of emergent beam
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 130.00
    SFr. 130.00
    1 * 1 ST
       SFr. 130.00
  • DIDAPOD060220
    Line generator, with a laser, enables to project one line, by the use of one glass cylinder, rotating at 360°
    Line generator, with a laser, enables to project one line, by the use of one glass cylinder, rotating at 360°
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 79.00
    SFr. 79.00
    1 * 1 ST
       SFr. 79.00
  • DIDAPOD010021
    Neutral density filter, d=0,9, set in an M49 ring
    Neutral density filter, d=0,9, set in an M49 ring
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 26.60
    SFr. 26.60
    1 * 1 ST
       SFr. 26.60
  • DIDAPOD010022
    Neutral density filter, d=3, set in an M49 ring
    Neutral density filter, d=3, set in an M49 ring
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 40.80
    SFr. 40.80
    1 * 1 ST
       SFr. 40.80
  • DIDAPOD010024
    Polariser, calibrated, set in an M49 ring
    Polariser, calibrated, set in an M49 ring
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 44.90
    SFr. 44.90
    1 * 1 ST
       SFr. 44.90
  • DIDAPOD010025
    Set of filters for Michelson interferometry for use with CCD Caliens camera
    Set of filters for Michelson interferometry for use with CCD Caliens camera
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 155.00
    SFr. 155.00
    1 * 1 ST
       SFr. 155.00
  • DIDAPOM052310
    Manual spectroscope, metal, lens focusing by extension of device, slot width adjustment by knurled ring.
    Manual spectroscope, metal, lens focusing by extension of device, slot width adjustment by knurled ring.
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 49.90
    SFr. 49.90
    1 * 1 ST
       SFr. 49.90
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Permanent frame, Ø 40 mm
    Permanent frame, Ø 40 mm
    DIDAPOD010095
    Log in per disponibilità
    1 * 1 ST - SFr. 40.80
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Simple lens holder, Ø 40 mm
    Simple lens holder, Ø 40 mm
    DIDAPOD010110
    Log in per disponibilità
    1 * 1 ST - SFr. 20.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Lens holder for fragile components, Ø 80 mm
    Lens holder for fragile components, Ø 80 mm
    DIDAPOD010080
    Log in per disponibilità
    1 * 1 ST - SFr. 71.50
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Lens holder, Ø 40/42 mm
    Lens holder, Ø 40/42 mm
    DIDAPOD010090
    Log in per disponibilità
    1 * 1 ST - SFr. 37.50
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Lens holder, Ø 80 mm
    Lens holder, Ø 80 mm
    DIDAPOD060130
    Log in per disponibilità
    1 * 1 ST - SFr. 57.40
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Stand, half moon, for optical components positioned remote from the Optical bench, height 210 mm
    Stand, half moon, for optical components positioned remote from the Optical bench, height 210 mm
    DIDAPOD002192
    Log in per disponibilità
    1 * 1 ST - SFr. 120.00