Orders LinkContactUsLinkComponent
Chlorine standard solution
 :   Hach
Chlorine standard solution
 :  Hach
Chlorine standard solution
Chlorine standard solution
For accuracy checks in chlorine determinations. Lot specific concentration displayed on label.
  • Simple accuracy checks
  • Minimise waste
  • Helps ensure reliable result
Product Family Options
  • Descrizione
    Concentrazione
    Conf
  • Chlorine standard solution, 2 ml PourRite ampules (NIST)
    25 - 30 mg/l as Cl₂
    20
    1 * 20 ST
    1 * 20 ST - 134.00 SFr.
    134.00 SFr.
    1 * 20 ST
       134.00 SFr.
  • Chlorine standard solution, 10 ml Voluette ampoules
    50 - 75 mg/l as Cl₂
    16
    1 * 16 ST
    1 * 16 ST - 201.00 SFr.
    201.00 SFr.
    1 * 16 ST
       201.00 SFr.
  • Chlorine standard solution, 2 ml PourRite ampules (NIST)
    50 - 75 mg/l as Cl₂
    20
    1 * 20 ST
    1 * 20 ST - 134.00 SFr.
    134.00 SFr.
    1 * 20 ST
       134.00 SFr.