Fleece hat, with Insulatex™ lining, HA10
Supplier: Portwest
Certificates
About this item
This classic fleece hat made of 100% polyester fleece is ideal for outdoor use.
- Soft fleece outer has high warmth to weight ratio
- Insulatex™ soft inner lining protects the head in harsh weather conditions
- When brim is unrolled the HA10 can be worn under a helmet
Weight: 280 g/m²
Specifications
- Catalog No:
- PRTWHA10BKR
- PRTWHA10ORR
- PRTWHA10YER
- PRTWHA10NAR
- Modello:
- HA10
- HA10
- HA10
- HA10
- Applicazioni per abiti:
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Tessuto:
- Polyester
- Polyester
- Polyester
- Polyester
- Peso:
- 280 g/m²
- 280 g/m²
- 280 g/m²
- 280 g/m²