Flacone da laboratorio, con filettatura, senza tappo a vite, DURAN®
Catalog # 215-3262
Supplier: DWK Life Sciences
Specifications
- Filettatura:GL 45
- Capacità:1500 ml
- Conf:1
Specifications
About this item
Manufactured from corrosion resistant AISI Type 316 L (1.4404) stainless steel, and hygienically constructed with all welded seams.
- Hazardous materials laboratory container for storage of liquids, intermediates, or solid product
- Storage of precious materials, such as high purity fine chemicals, pharmaceutical or cosmetic products
- Storage of light sensitive materials
The unbreakable steel bottle is ideal for storage applications where glass is not applicable, due to the risk of breakage or chemical incompatibility. The bottle has a smooth inner surface finish that corresponds to IIIc (DIN 17441) with rounded inner edges for ease of cleaning. Polished and brushed durable exterior finish. Bottle has a GL 45 thread with a built-in pouring lip and a rim for tapmer-evident caps. Supplied without a cap, stainless steel caps are available. It is fully compatible with all the DURAN Group GL 45 caps.
With printed retrace code (batch identification).