Orders LinkContactUsLinkComponent
BOD seed inoculum, POLYSEED
BOD seed inoculum, POLYSEED
BOD seed inoculum, POLYSEED, 50
Catalog # HACH2918700
Supplier:   Hach
BOD seed inoculum, POLYSEED
Catalog # HACH2918700
Supplier:  Hach
CAS Number:  
BOD seed inoculum, POLYSEED
BOD seed inoculum, POLYSEED
BOD seed inoculum, POLYSEED, 50
CAS Number:  
Price
SFr. 674.00
Log in for availability
1 * 50 ST
Product Details & Documents
BOD seed inoculum for use in biochemical oxygen demand (BOD5) test procedures. A blend of specialised microbial cultures in an easy-to-use capsule. Designed to provide a uniform standard for the degradation of both industrial and municipal waste in BOD5 analyses.
Ordering information: Package of 50 capsules. International Laboratory Supply (InterLab), LTD.
Documents
Specifications
Specifications
  • Conf:
    50
  • Descrizione:
    BOD seed inoculum, POLYSEED