Flame-retardant rain trousers, high visibility, Tielson 5874
Supplier: SIOEN
Certificates
About this item
These lightweight trousers feature a water repellent fabric on the outside and a moisture attracting coating on the inside. Made of 100% polyester fabric with 100% FR PU coating + AST.
- High tear resistance
- Fixed flame retardant cotton lining
- Leg narrowing by press studs
- Elasticated waist
- Taped seams
- Flame retardant reflective tape (50 mm)
The trousers are water- and windproof, anti-static and highly breathable.
Weight: 250 g/m²
Certifications: EN 343 (3-3), EN 471 (1-2), EN 1149-5, EN 13034 (PB/6), EN 531 (A B2 C1), EN ISO 14116 (inside: index 3; outside: index 1)
Specifications
- Catalog No:
- SIOE5874/278/L
- SIOE5874/278/3XL
- SIOE5874/278/2XL
- SIOE5874/279/S
- SIOE5874/278/1XL
- SIOE5874/279/M
- SIOE5874/279/L
- SIOE5874/279/3XL
- SIOE5874/279/2XL
- SIOE5874/279/1XL
- SIOE5874/278/S
- SIOE5874/278/M
- Modello:
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Tielson 5874
- Applicazioni per abiti:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Risk area:
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Rain (waterproof), Flame-retardant, Anti-static
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tessuto:
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- 100% polyester fabric with 100% FR PU coating
- Peso:
- 240 g/m²
- 240 g/m²
- 240 g/m²
- 240 g/m²
- 240 g/m²
- 240 g/m²
- 240 g/m²
- 240 g/m²
- 240 g/m²
- 240 g/m²
- 240 g/m²
- 240 g/m²