Orders LinkContactUsLinkComponent
Wheaton® MicroLiter™ plate sampling system
Wheaton® MicroLiter™ plate sampling system
Nome del fornitore:   MICROLITER ANALYTICAL SUPPLIES
Wheaton® MicroLiter™ plate sampling system
Nome del fornitore:  MICROLITER ANALYTICAL SUPPLIES
Certificates
Wheaton® MicroLiter™ plate sampling system
Wheaton® MicroLiter™ plate sampling system
5 varianti
Product Details & Documents
A comprehensive range of microplates, glass inserts, sealing mats and/or caps with chromatography-based septa that offer security for even the most volatile samples.
Polypropylene μLplates™ have a variety of closures that will stabilise the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. There is the option of choosing the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system permits even GC samples can be converted to 96-well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/silicone rubber using a mechanical seal. A variety of insert options are available for the appropriate application.
96-well round and square well microplates are available in a variety of volumes and options. Made of natural PP, they are made to ANSI specifications for 96-well microplates.
96-well round and square well covers are made of Ethylene Vinyl Acetate (EVA) and are pierceable by standard needle configurations found on most HPLC, LC/MS, CombiChem, and HTS instruments. They have a superior seal compared to similar products. For users who require some level of resealability, two silicone mats with a sprayed-on fluoropolymer resin barrier are available. Silicone mats are available in standard and pre-cut round or square well formats.
Product Family Options
Glass inserts
Informazioni sul prodotto
  • Volume pozzetti
    Disponibilità
    Price
  • MLAS07-0000-2200
    Glass inserts, 2,2 ml, clear glass, round, for 96 deep well μLplate® (2000 µl)
    Glass inserts, 2,2 ml, clear glass, round, for 96 deep well μLplate® (2000 µl)
    Log in per disponibilità
    Log in per disponibilità
    1 * 100 ST
    1 * 100 ST
µLplate® with glass inserts installed
Informazioni sul prodotto
  • Volume pozzetti
    Disponibilità
    Price
  • MLAS07-8000-1200
    One 96-well μLplate® (PP, deep well) with 1200 µl clear, round glass inserts (pre-inserted)
    One 96-well μLplate® (PP, deep well) with 1200 µl clear, round glass inserts (pre-inserted)
    2000 µl
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • MLAS07-8000-2200
    One 96-well μLplate® (PP, deep well) with 2200 µl clear, round glass inserts (pre-inserted)
    One 96-well μLplate® (PP, deep well) with 2200 µl clear, round glass inserts (pre-inserted)
    2000 µl
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
µLplate® with glass inserts installed and a detached μLmat™
Informazioni sul prodotto
  • Volume pozzetti
    Disponibilità
    Price
  • MLAS078020MB1200
    One 96-well μLplate® (PP, deep well) with 1200 µl clear, round glass inserts (pre-inserted) and a detached blue μLmat™ with pre-cut FEP/silicone septa
    One 96-well μLplate® (PP, deep well) with 1200 µl clear, round glass inserts (pre-inserted) and a detached blue μLmat™ with pre-cut FEP/silicone septa
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • MLAS078020MB2200
    One 96-well μLplate® (PP, deep well) with 2200 µl clear, round glass inserts (pre-inserted) and a detached blue μLmat™ with pre-cut FEP/silicone septa
    One 96-well μLplate® (PP, deep well) with 2200 µl clear, round glass inserts (pre-inserted) and a detached blue μLmat™ with pre-cut FEP/silicone septa
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
Snap caps and μLMATS™
Informazioni sul prodotto
  • Codice catalogo
    Descrizione
    Disponibilità
    Price
  • μLmat™, blue, with pre-cut FEP/silicone septa
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 0.00
    SFr. 0.00
    1 * 1 ST
       SFr. 0.00
Informazioni sul prodotto
  • Codice catalogo
    Descrizione
    Disponibilità
    Price
  • Snap cap, blue, with pre-cut FEP/silicone septa
    Log in per disponibilità
    Log in per disponibilità
    1 * 100 ST
    1 * 100 ST - SFr. 0.00
    SFr. 0.00
    1 * 100 ST
       SFr. 0.00
Covers and silicone mats
Informazioni sul prodotto
  • Codice catalogo
    Descrizione
    Disponibilità
    Price
  • EVA plate cover, natural, pierceable, for square well μLplates®
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 0.00