Boot covers, 440
bianco
Codice catalogo MMMA440
Nome del fornitore: 3M
Vai a
Product Details & Documents
These boot covers are made of combined SMS laminate and provide protection agaoinst non hazardous liquids and dust.
- Anti-static treated
- With ties for an excellent fit
- Serged seams
Length: 406 mm.
: Cat. I.
Documents
TDS Overboot with ties white 1 size
Scheda dati
Specifiche
- Colore:bianco
- Cat. No.:MMMA440
- Applicazioni per abiti:Laboratory