Cold resistant gloves, Cama Iso® 690+
PVC gloves with seamless fleece inlay, fully coated cuffs and roughened surface. The gloves provide short-term protection against temperatures down to −20 °C.
- High wearing comfort thanks to fleece inlay
- Very durable, good dexterity, high mechanical resistance
- Roughened surface for sure grip when handling wet parts
- Waterproof
- Cone-shaped cuff design
Ideal for winter use by local, district, regional and national services, drilling platforms, working and operating machines in cold areas, gardening and agriculture and for the preparation of ski-slopes.
Thickness: 3,5 ± 0,3 mm
Colour: Reddish brown
EN 388 (3221), EN 511 (121); Cat. II
: EN 388:2016 (3221X), EN 511 (021); Cat. II
- Catalog No:
- 112-1945
- 112-1946
- Modello:
- 690+
- 690+
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- Materiale rivestimento interno:
- maglia di lana
- maglia di lana
- Finitura:
- Rough
- Rough
- Tipo polsino:
- Svasato
- Svasato
- Norma EN:
- EN 388:2016 (3221X), EN 511 (021)
- EN 388:2016 (3221X), EN 511 (021)
Frequently Bought Together