Handheld refractometer, OE 200
Handheld refractometer, OE 200, 0 - 200 Oe° ; 0 - 45 °Bx ; 0 - 40 KI°, 1,3330 - 1,4100 nD
Codice catalogo 635-2115
Nome del fornitore: REICHERT
Vai a
Product Details & Documents
This handheld refractometer for wine provides fast and accurate measurements of dissolved sugars when testing grapes, citrus fruits, fruit juices, soft drinks, melons and maple sap for the winemaking industry.
Specifiche
- Modello:OE 200
- Range di misura:0 - 200 Oe° ; 0 - 45 °Bx ; 0 - 40 KI°
- Descrizione:Handheld refractometer
- Indice di rifrazione:1,3330 - 1,4100 nD
- Cat. No.:635-2115
- Bilance:Oechsle (Oeo), Brix (% sucrose) and Klosterneuburger (Klo)
- Range di compensazione della temperatura:10...40 °C