Handheld refractometers, Rhino
Supplier: REICHERT
Certificates
About this item
These automatic temperature-compensated (ATC) handheld refractometers are read directly and instantly.
- Water resistant and dustproof polycarbonate design for demanding environments
Brix scale based on percent by weight of sucrose allow for monitoring sugar level of soft drinks and juices, as well as to check dissolved solids in ripening fruit crops.
Specifications
- Catalog No:
- 635-2131
- 635-2130
- 635-2132
- Range di compensazione della temperatura:
- −18…+40 °C
- −18…+40 °C
- −18…+40 °C
- L×P×A:
- 235×38×38 mm
- 235×38×38 mm
- 235×38×38 mm
- Peso:
- 162,4 g
- 162,4 g
- 162,4 g