Orders LinkContactUsLinkComponent
Test strips
Test strips
Supplier:   Hach
Test strips
Supplier:  Hach
Certificates
Test strips
4 Options Available Below
Product Details & Documents
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
  • Portable - ideal for field testing of surface water
  • Easy to use
Documents
Product Family Options
Product Information
  • Descrizione
    Range di misura
    Tipo
    Conf
    Availability
    Price
  • Alkalinity (free and total CaCO₃)
    0 - 240 mg/L
    TS
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 51.20
    SFr. 51.20
    1 * 50 ST
       SFr. 51.20
  • Chloride
    30 - 600  mg/L
    TS
    40
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - SFr. 74.50
    SFr. 74.50
    1 * 40 ST
       SFr. 74.50
  • Chloride
    300 - 6000 mg/L
    TS
    40
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - SFr. 74.50
    SFr. 74.50
    1 * 40 ST
       SFr. 74.50
  • pH
    pH 4 - 9
    TS
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 27.80
    SFr. 27.80
    1 * 50 ST
       SFr. 27.80