Silicone rubber sponge stoppers
Marketsource
Nome del fornitore: Roth Carl
Vai a
Informazioni su questo articolo
These silicone rubber sponge stoppers are ideal for bacteriology, for long-termcultures, for filling and sterilising culture media and for the cultivating aerobic micro-organisms. Allows half the evaporation associated with regular cotton stoppers.
- Can be sterilised approx. 300 times at 180 °C for 10 hours
- Not hygroscopic
- Porous
- Washable and reusable
- Flexible, for test tubes and flasks
3 varianti
Product Details & Documents
These silicone rubber sponge stoppers are ideal for bacteriology, for long-termcultures, for filling and sterilising culture media and for the cultivating aerobic micro-organisms. Allows half the evaporation associated with regular cotton stoppers.
- Can be sterilised approx. 300 times at 180 °C for 10 hours
- Not hygroscopic
- Porous
- Washable and reusable
- Flexible, for test tubes and flasks
Product Family Options
Informazioni sul prodotto
- N. di foriAltezzaØ inferioreØ superioreDisponibilitàPrice
- Specifiche:MarketsourceMarketsource
- Specifiche:MarketsourceMarketsource
- Specifiche:MarketsourceMarketsource