Heat resistant gloves, Sekuroka®
Supplier: Roth Carl
Certificates
About this item
These highly flame and chemical resistant gloves are made of Nomex® polyamide fibres by DuPont. The finely-knitted jersey quality gloves are suitable for precision work.
- Long product life, good form stability, very comfortable fit
- Washable at max. +95 °C
- Protect against exposure to max contact heat of 100 °C (for max. 26 sec) and 250 °C (for max. 7,6 sec)
Certifications: EN 388, EN 407; Cat. II
Specifications
- Catalog No:
- ROTH0141.1
- ROTH0138.1
- ROTH0148.1
- ROTH0147.1
- ROTH0143.1
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- Tipo polsino:
- Dritto
- Dritto
- Dritto
- Dritto
- Dritto