Orders LinkContactUsLinkComponent
Covered sponge mops, PadMop™
Covered sponge mops, PadMop™
Nome del fornitore:   MICRONOVA
Covered sponge mops, PadMop™
Nome del fornitore:  MICRONOVA
Certificates
Covered sponge mops, PadMop™
Covered sponge mops, PadMop™
2 varianti
Product Details & Documents
Covered sponge mop features an angled head to apply the optimum amount of disinfectant in a pharmaceutical production or aseptic processing area. The low profile, brick-style mop and adapter create a fully padded mop head that handles large surface areas, as well as shelving, door jambs and corners. Absorbent foam interior retains liquid well. When pressure is applied, the moisture is released to fully saturate the surface to be disinfected. Launderable and autoclavable.
  • Dual padded cleaning surfaces for railings, door jambs, and corners
  • 19,05 mm urethane foam interiors
  • Holds up to common disinfectants and cleaning agents
  • Launderable and reusable
  • Autoclavable
Fully encased in a choice of low-particle fabric:
NovaPoly - 100% smooth knit polyester provides excellent disinfectant dispersal.
PolySorb - 100% textured polyester for removing residues and dispersing disinfectant.
The Micronova PadMop™ Covered Sponge Mop features dual cleaning surfaces for generous dispersement of solution on primary surfaces as well as using the top of the pad for cleaning door jambs, railings, piping, and more. The PadMop™ fits onto Micronova’s 360° swivel SlimLine™ adapter, or the LyoTool™ for cleaning lyophiliser shelving.
Informazioni per l'ordine: All components must be ordered separately. For additional handle lengths, see Universal Mop Handles or Plastic Mop Handles from Micronova.
Documents
Product Family Options
PadMop™
Informazioni sul prodotto
  • Dimensioni
    Disponibilità
    Price
  • 129-0744
    PadMop™ covered sponge mop, NovaPoly™, 100% smooth knit polyester, urethane foam interior
    PadMop™ covered sponge mop, NovaPoly™, 100% smooth knit polyester, urethane foam interior
    12.7×35 cm (5×14")
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 48.60
    SFr. 48.60
    1 * 1 ST
       SFr. 48.60
  • 129-0745
    PadMop™ covered sponge mop, NovaPoly™, irradiated, 100% smooth knit polyester, urethane foam interior
    PadMop™ covered sponge mop, NovaPoly™, irradiated, 100% smooth knit polyester, urethane foam interior
    12.7×35 cm (5×14")
    Log in per disponibilità
    Log in per disponibilità
    1 * 12 ST
    1 * 12 ST - SFr. 725.00
    SFr. 725.00
    1 * 12 ST
       SFr. 725.00
SlimLine™ Mop Adapters
Informazioni sul prodotto
  • Codice catalogo
    Descrizione
    Disponibilità
    Price
  • SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 543.00
    SFr. 543.00
    1 * 1 ST
       SFr. 543.00
Informazioni sul prodotto
  • Codice catalogo
    Descrizione
    Disponibilità
    Price
  • 18×4" (45.7×10 cm) mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 565.00
    SFr. 565.00
    1 * 1 ST
       SFr. 565.00
Stainless Steel Handles
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Descrizione
    Disponibilità
    Price
  • 1575 − 3048 mm
    Stainless steel quick disconnect extendable handle
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 557.00
    SFr. 557.00
    1 * 1 ST
       SFr. 557.00
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Descrizione
    Disponibilità
    Price
  • Acciaio inossidabile 304
    1295 cm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 200.00
    SFr. 200.00
    1 * 1 ST
       SFr. 200.00
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Descrizione
    Disponibilità
    Price
  • Acciaio inossidabile 304
    965 - 1829 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 449.00
    SFr. 449.00
    1 * 1 ST
       SFr. 449.00
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Descrizione
    Disponibilità
    Price
  • Fibra di vetro
    155 cm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 207.00
    SFr. 207.00
    1 * 1 ST
       SFr. 207.00
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Descrizione
    Disponibilità
    Price
  • Acciaio inossidabile 304
    203 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 142.00
    SFr. 142.00
    1 * 1 ST
       SFr. 142.00
Plastic Handles
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Descrizione
    Disponibilità
    Price
  • 1219 mm
    Plastic ACME threaded handle, with white hand grip
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 61.10
    SFr. 61.10
    1 * 1 ST
       SFr. 61.10
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Descrizione
    Disponibilità
    Price
  • 152 cm
    Plastic ACME threaded handle, with white hand grip
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 67.10
    SFr. 67.10
    1 * 1 ST
       SFr. 67.10
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Descrizione
    Disponibilità
    Price
  • 102 - 183 cm
    Plastic ACME threaded extendable handle, with white hand grip
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 206.00
    SFr. 206.00
    1 * 1 ST
       SFr. 206.00
Informazioni sul prodotto
  • Codice catalogo
    Lunghezza
    Descrizione
    Disponibilità
    Price
  • 1016 - 2565 mm
    Plastic ACME threaded extendable handle, with white hand grip
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 321.00
    SFr. 321.00
    1 * 1 ST
       SFr. 321.00