Tubing cutter for polymer tubing, Upchurch Scientific®
Codice catalogo 554-3024
Nome del fornitore: UPCHURCH SCIENTIFIC
Specifiche
- Descrizione:Standard polymer tubing cutter for 1/16" - 1/8" OD tubing, complete
- Conf:1
Specifiche
Informazioni su questo articolo
Designed to cut polymer tubing ¹/₁₆" and ¹/₈" O.D.
Complete with guide holes to ensure precise cutting. The cutter is durable, reliable and easy to operate.
Delivery: Complete unit supplied with five replacement blades.