Infusion filters set, Intrapur® Inline
Catalog # BRAU4099842N
Supplier: B. Braun
Specifications
- Membrana:0,2 µm
- Descrizione:Infusion sets for use with chemotherapeutic agents
- Conf:20
Specifications
About this item
Ready-to-use set with infusion filter.
- PVC free
- Retention of bacteria and particles
- High throughput
Specifically designed for use during chemotherapy.