Infusion valve, Safeflow®
Catalog # BRAU409100H
Supplier: B. Braun
Specifications
- Velocità del flusso:360 ml/min
- Volume:0,9 ml
- Colore:porpora/blu
- Descrizione:Infusion valves
- Conf:50
Specifications
About this item
This capless valve allows for a safe and convenient access during infusion therapy.
- Rapid delivery
- Compatible with luer-slip and -lock
- Provides needle-free IV access while eliminating the risk of needle stick injuries
Suitable for every IV procedure.