Staining jar according to Schiefferdecker
Staining jars acc. to Schiefferdecker, 90×70×50 mm
Codice catalogo KAVA632499890001
Nome del fornitore: Kavalier
Staining jar according to Schiefferdecker
Codice catalogo KAVA632499890001
Nome del fornitore: Kavalier
Vai a
Product Details & Documents
Made of soda-lime glass, with cover.
These staining trough hold up to 10 slides (76×26 mm), or 10 pairs of slides placed adjacently back-to-back.
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.
Specifiche
- Descrizione:Staining jars acc. to Schiefferdecker
- L×P×A:90×70×50 mm