Soxhlet extractors
Extractor with cone 29/32, 100 ml, 45/40 NS
Codice catalogo KAVA632442101100
Nome del fornitore: Kavalier
Vai a
Product Details & Documents
Borosilicate glass. For extraction from solids with solvents.
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.
Specifiche
- Contenuto estrattore:100 ml
- Cono femmina:45/40 NS
- Descrizione:Extractor with cone 29/32