Twisselmann extractors
Complete assembly, 250 ml, 29/32 NS
Codice catalogo KAVA632442108090
Nome del fornitore: Kavalier
Vai a
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Complete assembly for 250 ml flask size.
Specifiche
- Cono femmina:29/32 NS
- Descrizione:Complete assembly
- Volume matraccio:250 ml