Latex gloves
: DELTALAB
Disposable latex gloves designed for medical and general laboratory use.
- Powdered or powder-free
- Smooth finish
- Ambidextrous
- With beaded cuffs
Length: 240 mm
Thickness: 0,18 mm
Colour: Natural
EN 455, EN 374; AQL 1,5
: EN 455, EN 374; AQL 1,5
Disposable latex gloves designed for medical and general laboratory use.
- Powdered or powder-free
- Smooth finish
- Ambidextrous
- With beaded cuffs
Length: 240 mm
Thickness: 0,18 mm
Colour: Natural
EN 455, EN 374; AQL 1,5
: EN 455, EN 374; AQL 1,5
- Catalog No:
- DELT1000LM
- DELT1020LP
- Ambidestro/mano specifico:
- ambidestro
- ambidestro
- Monouso/riutilizzabile:
- monouso
- monouso
- Talcati/non talcati:
- Talcati
- Non talcati
- Finitura:
- Smooth
- Smooth
- Tipo polsino:
- Cuff_Style-21391099|Cuff_Style-21391103
- Cuff_Style-21391103|Cuff_Style-21391099
- Norma EN:
- EN 455, EN 374
- EN 455, EN 374
Product Family Options
- ModelloDimensione del guantoColoreLunghezzaSpessoreConfezione
- Powdered7 - 8naturale240 mm0,18 mm100/box1 * 100 STSFr. 25.40
Specifications
Cat. No.DELT1000LMAmbidestro/mano specificoambidestroMonouso/riutilizzabilemonousoTalcati/non talcatiTalcatiFinituraSmoothTipo polsinoCuff_Style-21391099|Cuff_Style-21391103Norma ENEN 455, EN 374 - Powder-free6 - 7naturale240 mm0,18 mm100/box1 * 1.000 STSFr. 185.00
Specifications
Cat. No.DELT1020LPAmbidestro/mano specificoambidestroMonouso/riutilizzabilemonousoTalcati/non talcatiNon talcatiFinituraSmoothTipo polsinoCuff_Style-21391103|Cuff_Style-21391099Norma ENEN 455, EN 374