D(+)-Talose
Bottiglia di vetro, 100 mg
Catalog # MOLE41482505-100MG
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: D-(+)-Talose, (2S,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal
- CAS Number:2595-98-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00135834
- Molecular Weight:180.16 g/mol
- EINECS:219-996-5
Specifications
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:100 mg
- Cat. No.:MOLE41482505-100MG
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



