Crucible for determination of volatile liquids
Catalog # KAVA641321098000
Supplier: Kavalier
Specifications
- Ø×A:32×42 mm
- Tipo:with lid, for determination of volatile liquid
- Capacità:22 ml
- Conf:1
Specifications
About this item
This crucible is made from porcelain.
Delivery: Supplied with lid.