Orders LinkContactUsLinkComponent
Assortment Boxes
Assortment Boxes
Supplier:   SEMADENI
Assortment Boxes
Supplier:  SEMADENI
Certificates
Assortment Boxes
3 Options Available Below
Product Details & Documents
Assortment and storage boxes, made of polystyrene, are crystal clear with hinged lid and without divisions.
Accessories information: The partitions allow storage boxes (330×220 mm) to be divided into convenience in many variants. Comes in different colours. The lids are transparent in colour.
Product Family Options
Product Information
  • Descrizione
    Colore
    L×P×A
    Availability
    Price
  • 250×170×45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 23.20
    SFr. 23.20
    1 * 1 ST
       SFr. 23.20
  • without lid
    Multicolore
    330×220×50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 27.60
    SFr. 27.60
    1 * 1 ST
       SFr. 27.60
  • 330×220×70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 29.10
    SFr. 29.10
    1 * 1 ST
       SFr. 29.10
Accessories
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0446
    Lids, PS, transparent, for compartments, 108×54 mm
    Log in for availability
    1 * 1 ST - SFr. 2.10
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0442
    Compartments, PS for storage boxes, yellow, 108×54 mm, 62 mm height
    Log in for availability
    1 * 1 ST - SFr. 3.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0443
    Compartments, PS for storage boxes, blue, 108×108 mm, 62 mm height
    Log in for availability
    1 * 1 ST - SFr. 5.10
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0445
    Lids, PS, transparent, for compartments, 54×54 mm
    Log in for availability
    1 * 1 ST - SFr. 1.70
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0436
    Compartments, PS for storage boxes, yellow, 108×54 mm, 45 mm height
    Log in for availability
    1 * 1 ST - SFr. 1.90
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0435
    Compartments, PS for storage boxes, red, 54×54 mm, 45 mm height
    Log in for availability
    1 * 1 ST - SFr. 1.60
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0444
    Compartments, PS for storage boxes, green, 162×108 mm, 62 mm height
    Log in for availability
    1 * 1 ST - SFr. 6.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0437
    Compartments, PS for storage boxes, blue, 108×108 mm, 45 mm height
    Log in for availability
    1 * 1 ST - SFr. 3.20
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0431
    Compartments, PS for storage boxes, red, 54×54 mm, 23 mm height
    Log in for availability
    1 * 1 ST - SFr. 1.60
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0441
    Compartments, PS for storage boxes, red, 54×54 mm, 62 mm height
    Log in for availability
    1 * 1 ST - SFr. 1.60
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0448
    Lids, PS, transparent, for compartments, 162×108 mm
    Log in for availability
    1 * 1 ST - SFr. 3.60
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0449
    Lids, PS, transparent, for compartments, 215×108 mm
    Log in for availability
    1 * 1 ST - SFr. 3.60
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • 211-2743
    Compartments, PS for storage boxes, green, 162×108 mm, 45 mm height
    Log in for availability
    1 * 1 ST - SFr. 5.10
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0439
    Compartments, PS for storage boxes, grey, 215×108 mm, 45 mm height
    Log in for availability
    1 * 1 ST - SFr. 5.80
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0447
    Lids, PS, transparent, for compartments, 108×108 mm
    Log in for availability
    1 * 1 ST - SFr. 3.10
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • SEMA0440
    Compartments, PS for storage boxes, grey, 322×108 mm, 45 mm height
    Log in for availability
    1 * 1 ST - SFr. 6.80