5-Fluoronicotinamide
Glass bottle for solids, 1 g
Catalog # ACTIAS23422G1
Marketsource
Supplier: ACTIVATE SCIENTIFIC
5-Fluoronicotinamide
Catalog # ACTIAS23422G1
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 70-58-6
Jump to:
Product Details & Documents
- CAS Number:70-58-6
- Formula:C₆H₅FN₂O
- MDL Number:MFCD00160352
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



