Antimonio standard dissolved in oil, 5,000 µg/g Sb in mineral oil Specpure® for wear metal analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
Physical form: liquid
MDL No.: MFCD00134030
Specifications
- N. MDL:MFCD00134030