ContactUsLinkComponent
Tubing connectors, L- shaped
  SEMA0545
 :  SEMADENI
undefined
Tubing connectors, L- shaped
  SEMA0545
 :  SEMADENI
  • Ø foro:
    2,5 mm
  • Ø int.tubo:
    4 - 5 mm
  • Conf:
    1
Made of PP.
  • Very good chemical resistance
  • Very good temperature resistance