Orders LinkContactUsLinkComponent
Empty solid phase extraction columns, CHROMABOND® | Avantor
Empty solid phase extraction columns, CHROMABOND®
Supplier:   MACHEREY-NAGEL
Empty solid phase extraction columns, CHROMABOND®
Supplier:  MACHEREY-NAGEL
Certificates
Empty solid phase extraction columns, CHROMABOND®
Empty solid phase extraction columns, CHROMABOND®
14 Options Available Below
Product Details & Documents
Empty and reservoir columns with accessories for solid phase extraction.
  • Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
  • Reservoir columns from PP for application of medium-size samples
  • Accessories for SPE columns (filter elements, adapters)
Product Family Options
CHROMABOND® empty columns
Product Information
  • Descrizione
    Capacità
    Availability
    Price
  • Empty PP columns with PE frits, one filter element is already inserted in the PP column
    1 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 223.00
    SFr. 223.00
    1 * 100 ST
       SFr. 223.00
  • Empty PP columns with PE frits, one filter element is already inserted in the PP column
    3 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 144.00
    SFr. 144.00
    1 * 50 ST
       SFr. 144.00
  • Empty PP columns with PE frits, one filter element is already inserted in the PP column
    6 ml
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - SFr. 98.90
    SFr. 98.90
    1 * 30 ST
       SFr. 98.90
  • Empty PP columns with PE frits, one filter element is already inserted in the PP column
    15 ml
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 117.00
    SFr. 117.00
    1 * 20 ST
       SFr. 117.00
  • Empty PP columns with PE frits, one filter element is already inserted in the PP column
    30 ml
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 124.00
    SFr. 124.00
    1 * 20 ST
       SFr. 124.00
  • Empty PP columns with PE frits, one filter element is already inserted in the PP column
    45 ml
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 162.00
    SFr. 162.00
    1 * 20 ST
       SFr. 162.00
  • Empty PP columns with PE frits, one filter element is already inserted in the PP column
    70 ml
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 175.00
    SFr. 175.00
    1 * 20 ST
       SFr. 175.00
  • Empty PP columns with PE frits, one filter element is already inserted in the PP column
    150 ml
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 192.00
    SFr. 192.00
    1 * 20 ST
       SFr. 192.00
  • Empty glass columns with glass fiber frits, one filter element is already inserted in the glass column
    3 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 284.00
    SFr. 284.00
    1 * 50 ST
       SFr. 284.00
  • Empty glass columns with glass fiber frits, one filter element is already inserted in the glass column
    6 ml
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - SFr. 223.00
    SFr. 223.00
    1 * 30 ST
       SFr. 223.00
  • Empty LV PP columns with PE frits, for 200/500 mg adsorbent weight
    15 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 225.00
    SFr. 225.00
    1 * 50 ST
       SFr. 225.00
Reservoir columns for application of medium-size samples
Product Information
  • Descrizione
    Capacità
    Availability
    Price
  • Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns
    30 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 127.00
    SFr. 127.00
    1 * 10 ST
       SFr. 127.00
  • Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns
    70 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 135.00
    SFr. 135.00
    1 * 10 ST
       SFr. 135.00
  • Reservoir columns, PP, with one adapter for 15, 45, 70 ml CHROMABOND® PP columns
    70 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 185.00
    SFr. 185.00
    1 * 10 ST
       SFr. 185.00
Accessories
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA732019
    PE frits for LV PP columns 15 ml, for 100 mg adsorbent weight
    Log in for availability
    1 * 250 ST - 147.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA732020
    PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight
    Log in for availability
    1 * 250 ST - 206.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730101
    Adapters (PP) for PP columns (1, 3 and 6 ml)
    Log in for availability
    1 * 10 ST - 107.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730105
    Adapters (PVDF) for glass columns (3 and 6 ml)
    Log in for availability
    1 * 10 ST - 223.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730026
    PE frits for PP columns, 70 ml
    Log in for availability
    1 * 250 ST - 313.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730192
    Glass fiber frits for glass columns, 6 ml
    Log in for availability
    1 * 250 ST - 265.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730351
    PE frits for PP columns, 15 ml
    Log in for availability
    1 * 250 ST - 265.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730356
    PE frits for PP columns, 45 ml
    Log in for availability
    1 * 250 ST - 304.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730475
    PE frits for PP columns, 150 ml
    Log in for availability
    1 * 250 ST - 336.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730034
    PE frits for PP columns, 30 ml
    Log in for availability
    1 * 250 ST - 287.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730566
    Adapters (PE) for PP columns (30 and 70 ml)
    Log in for availability
    1 * 1 KIT - 32.40 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730385
    Adapters (PE) for PP columns (15, 45 and 70 ml)
    Log in for availability
    1 * 10 ST - 139.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730191
    Glass fiber frits for glass columns, 3 ml
    Log in for availability
    1 * 250 ST - 218.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730164
    PE frits for PP columns, 1 ml
    Log in for availability
    1 * 250 ST - 145.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730163
    PE frits for PP columns, 6 ml
    Log in for availability
    1 * 250 ST - 252.00 SFr.
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • MANA730162
    PE frits for PP columns, 3 ml
    Log in for availability
    1 * 250 ST - 205.00 SFr.