Disposable earplugs, uvex hi-com detec
UVEX2112.114
: UVEX
- Colore:blu
- Descrizione:Detectable earplugs, with cord, packed in box
- Conf:100 PAIR
The hi-com detec is a detectable disposable earplug for the food industry. The innovative shape of the hi-com reduces the level of sound that can build up in the inner ear when wearing hearing protection. There is no risk of overprotection in areas with medium noise exposure.
- Earplug design make it possible to understand speech therefore improving communication
- Easy to insert, even in narrower ear canals
- Metal powder in cord allows full detection, even if the cord is shredded
- Blue colour enables easy recognition in production areas
SNR: 24 dB
: EN 352-2:2002