Weighing bottles, PP
Catalog # VITL80345
Supplier: VITLAB
Specifications
- Ø×A:30×30 mm
- Capacità:50 ml
- Conf:10
Specifications
About this item
Made of PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
Cylindrical shape, with knobbed lid.