Sealent wipes, KIMTECH PREP*
bianco, 400×500 mm, 500 sheets/pack
Codice catalogo 115-1556
Nome del fornitore: KIMBERLY CLARK
Vai a
Product Details & Documents
These soft and flexible wipes have a non woven open structure that is ideal for the removal of excess sealant prior to industrial painting or coating of a surface.
- Textured open weave material traps and removes excess sealant
- Pliable material helps remove sealant from hard to get to spaces
- Low linting performance helps minimise the occurrence of defects and cross-contamination from particulate matter
Specifiche
- Confezione:500 sheets/pack
- Colore:bianco
- L×L:400×500 mm