Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ KappaSelect
Affinity chromatography columns, HiTrap™ KappaSelect
Nome del fornitore:   Cytiva
Affinity chromatography columns, HiTrap™ KappaSelect
Nome del fornitore:  Cytiva
Certificates
Affinity chromatography columns, HiTrap™ KappaSelect
Affinity chromatography columns, HiTrap™ KappaSelect
2 varianti
Product Details & Documents
KappaSelect is an affinity medium designed for the purification of human Fab (kappa) fragments.
Antibody fragments, especially Fabs, are getting increased attention as potential biopharmaceuticals because they have some advantages over monoclonal antibodies (MAbs).
Specifications for Product Family
Specifiche
  • Catalog No:
  • 17-5458-12
  • 17-5458-11
  • Stoccaggio:
  • 4 to 8°C, 20% Ethanol
  • 4 to 8°C, 20% Ethanol
Specifiche
Product Family Options
Informazioni sul prodotto
  • Colonna
    Dimensione delle sfere
    Capacità
    Disponibilità
    Price
  • 17-5458-12
    HiTrap™ KappaSelect
    HiTrap™ KappaSelect
    PP
    16×25 mm
    5 ml
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 1’020.00
    SFr. 1’020.00
    1 * 1 ST
       SFr. 1’020.00
    Specifiche:
    More
    Specifications
    Cat. No.
    17-5458-12
    Stoccaggio
    4 to 8°C, 20% Ethanol
  • 17-5458-11
    HiTrap™ KappaSelect
    HiTrap™ KappaSelect
    PP
    7×25 mm
    1 ml
    Log in per disponibilità
    Log in per disponibilità
    1 * 5 ST
    1 * 5 ST - SFr. 1’070.00
    SFr. 1’070.00
    1 * 5 ST
       SFr. 1’070.00
    Specifiche:
    More
    Specifications
    Cat. No.
    17-5458-11
    Stoccaggio
    4 to 8°C, 20% Ethanol