High visibility rain jacket, Windsor 708Z
Informazioni su questo articolo
This waterproof and windproof jacket is made of Siopor® Extra fabric consisting of 100% PU laminated polyester fabric (cotton touch). The fabric is water repellent on the outside and moisture attracting on the inside. With fixed mesh lining in body and polyamide lining in sleeves.
- Straight collar, detachable hood with chin protection and peak
- Zip closure with long zip puller under flap with touch and close fastening
- Two inset chest pockets and two inset pockets with zip closure, sleeve pocket, Napoleon pocket
- Shaped sleeves, elasticated end with narrowing by touch and close fastening
- Underarm ventilation, taped seams
- Drawstring in waist and hem
Back length: 85 cm
Weight: 205 g/m²
Certifications: EN 471 (3-2), EN 343 (3-3)
12 varianti
Product Details & Documents
This waterproof and windproof jacket is made of Siopor® Extra fabric consisting of 100% PU laminated polyester fabric (cotton touch). The fabric is water repellent on the outside and moisture attracting on the inside. With fixed mesh lining in body and polyamide lining in sleeves.
- Straight collar, detachable hood with chin protection and peak
- Zip closure with long zip puller under flap with touch and close fastening
- Two inset chest pockets and two inset pockets with zip closure, sleeve pocket, Napoleon pocket
- Shaped sleeves, elasticated end with narrowing by touch and close fastening
- Underarm ventilation, taped seams
- Drawstring in waist and hem
Back length: 85 cm
Weight: 205 g/m²
Certifications: EN 471 (3-2), EN 343 (3-3)
Specifications for Product Family
Specifiche
- Catalog No:
- 113-7174
- 113-7173
- 113-7172
- 113-7169
- 113-7170
- 113-7171
- 113-7180
- 113-7179
- 113-7178
- 113-7175
- 113-7176
- 113-7177
- Modello:
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Windsor 708Z
- Applicazioni per abiti:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- 100% PU laminated polyester
- Peso:
- 205 g/m²
- 205 g/m²
- 205 g/m²
- 205 g/m²
- 205 g/m²
- 205 g/m²
- 205 g/m²
- 205 g/m²
- 205 g/m²
- 205 g/m²
- 205 g/m²
- 205 g/m²
Specifiche
Product Family Options
Informazioni sul prodotto
- ColoreTaglia dell'abbigliamentoDisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.113-7174ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7173ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7172ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7169ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7170ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7171ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7180ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7179ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7178ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7175ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7176ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m² - Specifiche:
More
Specifications
Cat. No.113-7177ModelloWindsor 708ZApplicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexTipo di chiusuraCon chiusura a zipTessuto100% PU laminated polyesterPeso205 g/m²