Orders LinkContactUsLinkComponent
Scaffolding lanyards, Barracuda™, Miller®
Scaffolding lanyards, Barracuda™, Miller®
Nome del fornitore:   HONEYWELL SAFETY
Scaffolding lanyards, Barracuda™, Miller®
Nome del fornitore:  HONEYWELL SAFETY
Certificates
Scaffolding lanyards, Barracuda™, Miller®
2 varianti
Product Details & Documents
These fall arrest rope lanyards with shock absorber, vertical scaffold clamp and screwgate karabiner provide an ideal fall protection solution for scaffolders. Barracuda™ is the only practical system that allows anchorage above waist height, thus significantly reducing the fall arrest forces from a fall and consequently the risk of injury.
  • Jaw system with clamp teeth, PP grip
  • Anchorage clamp is easily fitted using one hand thus allowing the scaffolder to keep one hand free
  • Anchorage point above waist height offers the scaffolder a greater working area
For use with scaffolding tubes from 48,3 to 50 mm.
Certifications: EN 795 Class B (clamp), EN 362-04 (connector)
Specifications for Product Family
Specifiche
  • Catalog No:
  • 111-1390
  • 111-1392
  • Modello:
  • 1,2 m
  • 2 m
  • Tensione:
  • > 2200 daN
  • > 2200 daN
  • Tipo di connettore:
  • Zinc finished steel
  • Galvanized steel
  • Peso:
  • 1,22 kg
  • 1,20 kg
Specifiche
Product Family Options
Informazioni sul prodotto
  • Lunghezza
    Disponibilità
    Price
  • 1,2 m
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 529.00
    SFr. 529.00
    1 * 1 ST
       SFr. 529.00
    Specifiche:
    More
    Specifications
    Cat. No.
    111-1390
    Modello
    1,2 m
    Tensione
    > 2200 daN
    Tipo di connettore
    Zinc finished steel
    Peso
    1,22 kg
  • 1,75 m
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 564.00
    SFr. 564.00
    1 * 1 ST
       SFr. 564.00
    Specifiche:
    More
    Specifications
    Cat. No.
    111-1392
    Modello
    2 m
    Tensione
    > 2200 daN
    Tipo di connettore
    Galvanized steel
    Peso
    1,20 kg