Burn cover
Burn covers, packed in box containing 10 sterile plasters (74×45 mm)
Codice catalogo 118-0384
Nome del fornitore: ORKLA CARE
Vai a
Product Details & Documents
This cover protects and relieves the pain caused by minor burns. The cushion is a cool water-based hydrogel that covers the nerve endings of the skin, helping to relieve pain. It also creates a moist environment that prevents scabs from forming and promotes faster healing.
- Blue colour - suitable for use when handling food
- Promotes faster healing
- Does not attach to wound
- Can also be used for minor cuts and grazes
- Box can be used in Cederroth’s first aid stations
Specifiche
- Descrizione:Burn covers, packed in box containing 10 sterile plasters (74×45 mm)