Orders LinkContactUsLinkComponent
Ultra fine absorption grains
Ultra fine absorption grains
Supplier:   asecos
Ultra fine absorption grains
Supplier:  asecos
Certificates
Ultra fine absorption grains
Ultra fine absorption grains
2 Options Available Below
Product Details & Documents
Fine absorption granules for oil, alkalis, grease, motor fuels, varnishes.
  • Officially tested and approved for use on traffic areas
  • 0,3 to 0,7 mm granularity
  • 10 kg sack binds up to 11 litres of heating oil
  • Fills smallest cracks and openings
  • Developed for special purposes where a minimum of waste and a maximum of absorption is needed
Product Family Options
Product Information
  • Descrizione
    Assorbanza
    Confezione
    Availability
    Price
  • Ultra fine absorption granules (0,3 - 0,7 mm)
    11 l/sack
    10 kg/sack
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 45.30
    SFr. 45.30
    1 * 1 ST
       SFr. 45.30
  • Ultra fine absorption granules (0,3 - 0,7 mm)
    11 l/sack
    3×10 kg/sack
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 112.00
    SFr. 112.00
    1 * 1 ST
       SFr. 112.00