Universal absorbent rolls
: asecos
Universal rolls for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
- Absorbency of a capacity up to 16 times of its own weight
- High rib and rub resistance
- Standard material for small leaks and prevention
- Heavyweight material for small leaks or the absortion of spillage
- Rolls are perforated to take sufficient material as required
Universal rolls for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
- Absorbency of a capacity up to 16 times of its own weight
- High rib and rub resistance
- Standard material for small leaks and prevention
- Heavyweight material for small leaks or the absortion of spillage
- Rolls are perforated to take sufficient material as required
Product Family Options
- DescrizioneConfezioneL×L
- Standard universal roll2 rolls/pack90 m×400 mm1 * 1 STSFr. 369.00
- Standard universal roll1 roll/pack90 m×800 mm1 * 1 STSFr. 369.00
- Heavyweight universal roll1 roll/pack45 m×400 mm1 * 1 STSFr. 369.00
- Heavyweight universal roll2 rolls/pack45 m×800 mm1 * 1 STSFr. 369.00
Accessories
- Descrizione
- Descrizione
- Descrizione
- MSGA9905Rack with cutting edge for rolls which need to be ready at hand, up to 40 cm width1 * 1 ST - SFr. 88.40