Orders LinkContactUsLinkComponent
Cuvettes and holders
Cuvettes and holders
Supplier:   BIOCHROM
Cuvettes and holders
Supplier:  BIOCHROM
Certificates
Cuvettes and holders
9 Options Available Below
Product Details & Documents
Cuvettes for spectrophotometers designed to hold samples during spectroscopic experiments.
Different pathlengths and formats are available.
Product Family Options
Cuvettes
Product Information
  • Descrizione
    Volume
    Cammino ottico
    Availability
    Price
  • Quartz flow through cell
    450 µl
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Glass flow through cell
    450 µl
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Semi micro glass cell
    800 µl
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Cuvette holders
Product Information
  • Descrizione
    Volume
    Cammino ottico
    Availability
    Price
  • Cell holder for up to 1 - 10 mm pathlength
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Cell holder for 10 - 50 mm pathlength cuvettes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Four-position cell changer, 10 - 50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microvolume cell holder for 50 μl cells
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Programmable Peltier heated cell holder (20 - 105 °C)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Ultra microvolume cell holder with two-axis adjustment for 5 μl cells