Tray, with lid
17 l, 430×331×195 mm
Codice catalogo 231-0408
Nome del fornitore: VITLAB
Vai a
Product Details & Documents
Rectangular tray, made of white PP with lid, ideal for a storage applications.
- Easy to clean
- Broad, stable, easy to grip edge
- Smooth surface, rounded corners and edges
Specifiche
- Capacità:17 l
- L×P×A:430×331×195 mm