Orders LinkContactUsLinkComponent
PIG® utility pans
PIG® utility pans
4 L, 270×270×80 mm, 12/box
Codice catalogo NEWPPAN404
Nome del fornitore:   NEW PIG
PIG® utility pans
Codice catalogo NEWPPAN404
Nome del fornitore:  NEW PIG
PIG® utility pans
PIG® utility pans
4 L, 270×270×80 mm, 12/box
Price
SFr. 55.20
Log in per disponibilità
1 * 12 ST
Product Details & Documents
Light-weight, economical Utility Pans serve dozens of purposes throughout your workplace.
  • Chemical-resistant polypropylene can be used with a variety of workplace materials
  • Nestable design occupies minimal space for shipping and storing
  • Cleans easily with soap and water
Capture drips, temporarily contain small parts, organize a workbench and much more.
Specifiche
  • Confezione:
    12/box
  • Capacità:
    4 L
  • L×P×A:
    270×270×80 mm