VWR®, Applicatore di pesi a piastra a contatto, Peso a contatto
Catalog # 710-0961
Supplier: VWR Collection
CAS Number:
Specifications
- Conf:1
- Descrizione:Applicatore per piastre a contatto, Contact Weight
Specifications
About this item
Used in combination with contact plates, the contact weight enables the standardisation of microbiological surface control between operators. It applies the same constant pressure for a pre-set time on a contact plate. This ensures results are directly comparable. The contact weight meets the ISO 18593 standard, and has a sterilisable stainless steel base and pull-out digital timer.
- Base in acciaio inox sterilizzabile a carole secco o a vapore con timer digitale estraibile
- Da utilizzare con diversi tipi di piastre a contatto
- A standard ISO 18593
- Peso di appena 500 g
Timer included provided without the certificate of calibration; if a calibration is required it is necessary to buy the article 609-0184 as accessory that includes the calibration.