ContactUsLinkComponent
Reaction/distillation unit, one-way
Reaction/distillation unit, one-way
  LENZ06265597
undefined
Reaction/distillation unit, one-way
  LENZ06265597
  • Descrizione:
    Reaction/distillation unit, one way, 30 l vessel, LF 200
  • Conf:
    1
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes