Orders LinkContactUsLinkComponent
Glutamic-C™, sequencing grade
Glutamic-C™, sequencing grade
SG-Glutamic-C™, 2×10 μg, 2
Codice catalogo 786-15
Nome del fornitore:   G-Biosciences
Glutamic-C™, sequencing grade
Codice catalogo 786-15
Nome del fornitore:  G-Biosciences
Numero CAS:  
Glutamic-C™, sequencing grade
SG-Glutamic-C™, 2×10 μg, 2
Numero CAS:  
Price
SFr. 354.00
Log in per disponibilità
1 * 2 ST
Product Details & Documents
SG-Glutamic-C™ is a serine endopeptidase, from S. aureus V8, that is highly specific for the cleavage of peptide bonds at the carboxy side of either aspartic or glutamic acid, depending on the buffer used. In Tris-HCl buffer, in particular in the absence of phosphate ions, the enzyme is specific for the glutamyl site. Recommended buffers for fragmentation of proteins using this enzyme are 50 mM Tris-HCI, pH 8,0 or bicarbonate buffer. Highly purified preparations of SG-Glutamic-C™ are chemically modified making the enzyme both resistant to autolysis and stabilises its enzymatic activity.
  • Modified glutamic-C for sequence analysis
  • Resistant to autolysis and degradation
  • High specificity, free from other known endopeptidases
  • Consistency of activity levels from lot to lot for reproducible digestions
SG-Glutamic-C™ is supplied lyophilised in 10 μg vials. The enzyme is typically reconstituted to a concentration of 0,5 μg/ml and commonly used at a ratio of 1:100 to 1:20 (enzyme to protein, by weight) in a standard digestion buffer.
Documents
Specifiche
Specifiche
  • Conf:
    2
  • Descrizione:
    SG-Glutamic-C™, 2×10 μg
Specifiche
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.