Empty columns, HiScale™
Informazioni su questo articolo
HiScale™ empty chromatography columns are a family of pressure-stable, easy to pack, columns for medium to high pressure liquid chromatography using a chromatography system.
- Convenient column measurement scale, nonrotating plunger mechanism, and ergonomically designed components for easier adjustment during column packing
- Built to withstand pressures up to 20 bar (2 MPa, 290 psi) for compatibility with modern BioProcess™ resins
- Dual adapters and axial compression capability allow for a large range of bed heights and column packing protocols
- Choose from 10, 16, 26, or 50 mm inner diameter, and 200 mm or 400 mm length to accommodate bed volumes up to 785 ml
- Designed to facilitate robust process scaleup, as well as cost-efficient and accurate scaledown for applications such as viral clearance studies
- Equipped with two adapters allowing for a large range of bed heights
HiScale™ pressure‐stable, empty columns are designed for process development and preparative chromatography using ÄKTA protein purification systems. These chromatography columns withstand pressures up to 20 bar, with a user‐friendly design that supports process optimization through simplified and reproducible operations.
The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments as well as disassembly and cleaning. The entire shaft may be rinsed after column packing, minimizing the risk of bacterial growth. The column’s measurement scale helps users to predict and reach a predefined bed height.
HiScale™ columns are based on the well‐established line of XK columns. They are compatible with all methods currently applicable to XK columns.
HiScale™ 10/40 columns are especially suitable for scaledown studies used in, for example, viral clearance evaluation in downstream processing. The small inner diameter and flexible adapters minimize costly sample and viral spike volumes. The robust design of HiScale™ 10/40 allows for controlled bed heights and gives reproducible results offering an effective scaled down model of the large-scale downstream process.
HiScale™ chromatography columns are compatible with all Cytiva resins. For columns packed with SOURCE 15 and SOURCE 30, a 10 µm net ring should be used.
: Each HiScale™ column is delivered with a column tube, outer protection tube, two adapters, PEEK tubing, and unions at both ends for easy connection to valves, pumps, and monitors. For HiScale™ 10/40, connectors are provided in the accessory kit included in the package.
7 varianti
Product Details & Documents
HiScale™ empty chromatography columns are a family of pressure-stable, easy to pack, columns for medium to high pressure liquid chromatography using a chromatography system.
- Convenient column measurement scale, nonrotating plunger mechanism, and ergonomically designed components for easier adjustment during column packing
- Built to withstand pressures up to 20 bar (2 MPa, 290 psi) for compatibility with modern BioProcess™ resins
- Dual adapters and axial compression capability allow for a large range of bed heights and column packing protocols
- Choose from 10, 16, 26, or 50 mm inner diameter, and 200 mm or 400 mm length to accommodate bed volumes up to 785 ml
- Designed to facilitate robust process scaleup, as well as cost-efficient and accurate scaledown for applications such as viral clearance studies
- Equipped with two adapters allowing for a large range of bed heights
HiScale™ pressure‐stable, empty columns are designed for process development and preparative chromatography using ÄKTA protein purification systems. These chromatography columns withstand pressures up to 20 bar, with a user‐friendly design that supports process optimization through simplified and reproducible operations.
The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments as well as disassembly and cleaning. The entire shaft may be rinsed after column packing, minimizing the risk of bacterial growth. The column’s measurement scale helps users to predict and reach a predefined bed height.
HiScale™ columns are based on the well‐established line of XK columns. They are compatible with all methods currently applicable to XK columns.
HiScale™ 10/40 columns are especially suitable for scaledown studies used in, for example, viral clearance evaluation in downstream processing. The small inner diameter and flexible adapters minimize costly sample and viral spike volumes. The robust design of HiScale™ 10/40 allows for controlled bed heights and gives reproducible results offering an effective scaled down model of the large-scale downstream process.
HiScale™ chromatography columns are compatible with all Cytiva resins. For columns packed with SOURCE 15 and SOURCE 30, a 10 µm net ring should be used.
: Each HiScale™ column is delivered with a column tube, outer protection tube, two adapters, PEEK tubing, and unions at both ends for easy connection to valves, pumps, and monitors. For HiScale™ 10/40, connectors are provided in the accessory kit included in the package.
Specifications for Product Family
Specifiche
- Catalog No:
- 29-3605-50
- 28-9644-41
- 28-9644-24
- 28-9645-14
- 28-9645-13
- 28-9644-45
- 28-9644-44
- Pressione max.:
- 20 bar
- 20 bar
- 20 bar
- 20 bar
- 20 bar
- 20 bar
- 20 bar
- Limite di temperatura:
- 4...40 °C
- 4...40 °C
- 4...40 °C
- 4...40 °C
- 4...40 °C
- 4...40 °C
- 4...40 °C
Specifiche
Product Family Options
Informazioni sul prodotto
- ColonnaDimensione delle sfereDisponibilitàPrice
- 29-3605-50HiScale™ 10/40Specifiche:
More
Specifications
Cat. No.29-3605-50Pressione max.20 barLimite di temperatura4...40 °C - 28-9644-41HiScale™ 16/20Specifiche:
More
Specifications
Cat. No.28-9644-41Pressione max.20 barLimite di temperatura4...40 °C - 28-9644-24HiScale™ 16/40Specifiche:
More
Specifications
Cat. No.28-9644-24Pressione max.20 barLimite di temperatura4...40 °C - 28-9645-14HiScale™ 26/20Specifiche:
More
Specifications
Cat. No.28-9645-14Pressione max.20 barLimite di temperatura4...40 °C - 28-9645-13HiScale™ 26/40Specifiche:
More
Specifications
Cat. No.28-9645-13Pressione max.20 barLimite di temperatura4...40 °C - 28-9644-45HiScale™ 50/20Specifiche:
More
Specifications
Cat. No.28-9644-45Pressione max.20 barLimite di temperatura4...40 °C - 28-9644-44HiScale™ 50/40Specifiche:
More
Specifications
Cat. No.28-9644-44Pressione max.20 barLimite di temperatura4...40 °C
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Connector 1/16" male/Luer female, for HiScale™ 16 and 26Connector 1/16" male/Luer female, for HiScale™ 16 and 2618-1112-51Log in per disponibilità1 * 2 ST - SFr. 114.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Short column holder, for HiScale™ 16 and 26Short column holder, for HiScale™ 16 and 2618-1113-17Log in per disponibilità1 * 1 ST - SFr. 293.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Column clamp, OD 10 - 21 mm
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Column holder, steel, for HiScale™ 50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Packing tube 40, for HiScale™ 26
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Packing tube 40, for HiScale™ 50
Informazioni sul prodotto
- Codice catalogoMaterialeDisponibilitàPrice
- Spanner wrench, for nuts Ø 58 - 62 mm, for HiScale™ 16 columnsSpanner wrench, for nuts Ø 58 - 62 mm, for HiScale™ 16 columns28-9647-76Acciaio GalvanizzatoLog in per disponibilità1 * 2 ST - SFr. 95.20
Informazioni sul prodotto
- Codice catalogoMaterialeDisponibilitàPrice
- Spanner wrench, for nuts Ø 65 - 75 mm, for HiScale™ 26 columnsSpanner wrench, for nuts Ø 65 - 75 mm, for HiScale™ 26 columns28-9647-77Acciaio GalvanizzatoLog in per disponibilità1 * 2 ST - SFr. 106.00
Informazioni sul prodotto
- Codice catalogoMaterialeDisponibilitàPrice
- Spanner wrench, for nuts Ø 95 - 100 mm, for HiScale™ 50 columnsSpanner wrench, for nuts Ø 95 - 100 mm, for HiScale™ 50 columns28-9647-78Acciaio GalvanizzatoLog in per disponibilità1 * 2 ST - SFr. 117.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Union Valco F/F, PEEK, for HiScale™ 16 and 26Union Valco F/F, PEEK, for HiScale™ 16 and 2611-0003-39Log in per disponibilità1 * 5 ST - SFr. 220.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Fingertight stop plug, 1/16", PEEK, for HiScale™ 16 and 26Fingertight stop plug, 1/16", PEEK, for HiScale™ 16 and 2611-0003-55Log in per disponibilità1 * 5 ST - SFr. 83.90
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Superloop, M6 fittings, 150 ml, for HiScale™ 16, 26 and 50Superloop, M6 fittings, 150 ml, for HiScale™ 16, 26 and 5018-1023-85Log in per disponibilità1 * 1 ST - SFr. 1’810.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Stop plug 5/16" male, PEEK, for HiScale™ 50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Accessory kit for HiScale™ 16, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugsAccessory kit for HiScale™ 16, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs28-9663-67Log in per disponibilità1 * 1 KIT - SFr. 95.20
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Accessory kit for HiScale™ 26, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugsAccessory kit for HiScale™ 26, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs28-9663-74Log in per disponibilità1 * 1 KIT - SFr. 106.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Tubing, inner Ø 2 mm, for HiScale™ 50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Net ring, 20 μm, PP/PA (nylon), for HiScale™ 16Net ring, 20 μm, PP/PA (nylon), for HiScale™ 1628-9663-79Log in per disponibilità1 * 5 ST - SFr. 87.70
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Net ring, 20 μm, PP/PA (nylon), for HiScale™ 26Net ring, 20 μm, PP/PA (nylon), for HiScale™ 2628-9663-80Log in per disponibilità1 * 5 ST - SFr. 95.20
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Net ring, 20 μm, PP/PA (nylon), for HiScale™ 50Net ring, 20 μm, PP/PA (nylon), for HiScale™ 5028-9663-81Log in per disponibilità1 * 5 ST - SFr. 117.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Adapter, for HiScale™ 50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 16Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 1628-9666-46Log in per disponibilità1 * 1 ST - SFr. 581.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 26Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 2628-9666-48Log in per disponibilità1 * 1 ST - SFr. 983.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 50Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 5028-9666-49Log in per disponibilità1 * 1 ST - SFr. 1’280.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 50Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 5028-9666-50Log in per disponibilità1 * 1 ST - SFr. 1’920.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 26Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 2628-9666-51Log in per disponibilità1 * 1 ST - SFr. 1’250.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 16Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 1628-9666-52Log in per disponibilità1 * 1 ST - SFr. 754.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- O-ring, packing tube, for HiScale™ 16
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- O-ring, 26×4 mm, for HiScale™ 26
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- O-ring, 48×4 mm, for HiScale™ 50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Tubing, outer Ø 2 mm, cut, for HiScale™ 50Tubing, outer Ø 2 mm, cut, for HiScale™ 5028-9666-56Log in per disponibilità1 * 0,295 m - SFr. 95.20
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Packing tube 20, for HiScale™ 50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Packing tube 40, for HiScale™ 16
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Packing tube 20, for HiScale™ 16
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Packing tube 20, for HiScale™ 26
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Accessory kit for HiScale™ 50, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugsAccessory kit for HiScale™ 50, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs28-9663-75Log in per disponibilità1 * 1 KIT - SFr. 156.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Tubing, inner Ø 1 mm, for HiScale™ 16 and 26, beigeTubing, inner Ø 1 mm, for HiScale™ 16 and 26, beige18-1115-83Log in per disponibilità1 * 1 ST - SFr. 103.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Stop plug female, 1/16”