Informazioni su questo articolo
CM Sepharose™ Fast Flow is a weak cation exchanger based on the well established Sepharose™ Fast Flow ion exchange platform, extensively used for preparative protein separations in both research and industrial applications.
- Well proven weak anion exchanger developed for industrial downstream processes
- Used extensively for capture and intermediate purification of a wide range of approved biopharmaceuticals
- High chemical stability allows for well proven CIP and sanitisation protocols
- The hydrophilic nature of the base matrix ensures low levels of non specific binding leading to low levels of host cell-derived impurities in the elution pool
CM Sepharose Fast Flow has high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitisation protocols. Scale-up with CM Sepharose Fast Flow is straight forward and the medium is available in a range of pre-packed process development tools. As member of the BioProcess media range, CM Sepharose Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
CM Sepharose Fast Flow is part of the Sepharose Fast Flow ion exchange platform, which has been the industrial standard for ion exchange chromatography during recent decades. It is composed of crosslinked 6% agarose beads, with carboxymethyl (CM) weak cation exchange groups.
Specifiche
- Range di pH:4 - 13
- Matrice:Cross-linked agarose, 6%, spherical
- Ligando:Carboxymethyl
- Temperatura di conservazione:4 - 30 °C
- Capacità ionica totale:0,09 - 0,13 mmol H+/ml
Specifiche
Informazioni correlate
17071901_HAND BOOK 17071901_INSTRUCTIONS 17071901_INSTRUCTIONS 1 17071901_INSTRUCTIONS 2 17071901_INSTRUCTIONS 3
Manuale
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements