Magnetic stirring bar retriever, flexible, PTFE
VITL318597
: VITLAB
- Lunghezza:330 mm
- Conf:5
Flexible PTFE magnetic stirring bar retriever.
- High chemical resistance
- Magnet encapsulated (Ø×L: 12,5×51 mm)
- Easy to clean
Flexible PTFE magnetic stirring bar retriever.