Protease inhibitor cocktail, Plant ProteaseArrest™
Catalog # 733-1948
Supplier: G-Biosciences
CAS Number:
Specifications
- Conf:1 mL
- Descrizione:Plant ProteaseArrest™ (100X)
Specifications
About this item
A broad range, 100X concentrated, ready-to-use protease inhibitor cocktail, Plant ProteaseArrest™ inhibits plant serine, cysteine and other plant specific proteases including aminopeptidases, aspartic and metalloproteases.
- Plant specific protease inhibitor cocktail
- 100X concentrated, ready-to-use solution
Applications inhibition of protease activity in protein preparations of plant lysates and protection of proteins from proteolysis in such applications as electrophoresis, purification, storage and assays.